siRNA Details
Gene Symbol: Dexi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021428.4 | NP_067403.2 | 1 | 493 | ||
GenBank mRNA | AF152470.2 | - | 1 | 194 | ||
AK007644.1 | - | 1 | 494 | |||
AK032104.1 | - | 494 | ||||
AK183993.1 | - | 1 | 62 | |||
BC017551.1 | - | 1 | 480 | 4910742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
