siRNA Details
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021429.3 | NP_067404.2 | 2, 3 | 229 | ||
GenBank mRNA | AJ132192.1 | - | 2, 3 | 223 | ||
AK040654.1 | - | 2, 3 | 229 | |||
AK084802.1 | - | 2, 3 | 222 | |||
AK154696.1 | - | 2, 3 | 225 | |||
AK170838.1 | - | 2, 3 | 216 | |||
BC095977.1 | - | 2, 3 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
