siRNA Details
Gene Symbol: 1700123O20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021437.2 | NP_067412.1 | 2 | 1050 | ||
GenBank mRNA | AB041657.1 | - | 2 | 1005 | ||
AK007261.1 | - | 2 | 451 | |||
AK167989.1 | - | 2 | 234 | |||
AK170890.1 | - | 2 | 383 | |||
BC011283.1 | - | 2 | 179 | 3581864 |

More Information
Other Species Targeted

Gene Ontology Categories:
