siRNA Details
Gene Symbol: Eid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025613.3 | NP_079889.2 | 1 | 489 | ||
GenBank mRNA | AF509968.1 | - | 1 | 427 | ||
AK002559.1 | - | 1 | 495 | |||
AK005258.1 | - | 1 | 459 | |||
AK011293.1 | - | 1 | 481 | |||
AK012453.1 | - | 1 | 481 | |||
AK014440.1 | - | 1 | 435 | |||
AK017543.1 | - | 1 | 465 | |||
AK077783.1 | - | 1 | 469 | |||
AK078402.1 | - | 1 | 435 | |||
AK078409.1 | - | 1 | 435 | |||
AK078826.1 | - | 1 | 413 | |||
AK148555.1 | - | 1 | 430 | |||
AK169604.1 | - | 1 | 463 | |||
AK172612.1 | - | 1 | 270 | |||
AL357453.1 | - | 1 | 455 | |||
BC010712.1 | - | 1 | 474 | |||
BC069897.1 | - | 1 | 412 | |||
BY703074.1 | - | 1 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
