siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164325.1 | NP_001157797.1 | 29, 30 | 4839 | ||
NM_021450.2 | NP_067425.2 | 29, 30 | 4842 | |||
GenBank mRNA | AF149013.1 | - | 29, 30 | 4792 | ||
AF376052.1 | - | 29, 30 | 4695 | |||
AK029000.1 | - | 4, 5 | 829 | |||
AY032951.1 | - | 29, 30 | 4811 | |||
BC026440.1 | - | 4, 5 | 398 | |||
BC145122.1 | - | 29, 30 | 4587 | |||
BC150709.1 | - | 29, 30 | 4637 | 9087620 |
