siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164325.1 | NP_001157797.1 | 20 | 2949 | ||
NM_021450.2 | NP_067425.2 | 20 | 2949 | |||
GenBank mRNA | AF149013.1 | - | 20 | 2899 | ||
AF376052.1 | - | 20 | 2805 | |||
AK049329.1 | - | 5 | 851 | |||
AY032951.1 | - | 20 | 2918 | |||
BC145122.1 | - | 20 | 2744 | |||
BC150709.1 | - | 20 | 2744 | 9087620 |
