siRNA Details
Gene Symbol: Pmaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021451.2 | NP_067426.1 | 2 | 248 | ||
GenBank mRNA | AB041230.1 | - | 2 | 248 | ||
AK043856.1 | - | 2 | 279 | |||
AK088556.1 | - | 2 | 258 | |||
AK143990.1 | - | 2 | 320 | |||
AK169914.1 | - | 2 | 292 | |||
BC050821.1 | - | 2 | 235 | 6517820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
