siRNA Details
Gene Symbol: Mlxipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021455.4 | NP_067430.2 | 13 | 2051 | ||
GenBank mRNA | AF156604.1 | - | 13 | 2036 | ||
AF245475.1 | - | 13 | 2029 | |||
AF245476.1 | - | 13 | 2057 | |||
AF245477.1 | - | 14 | 1963 | |||
AF245478.1 | - | 13 | 2029 | |||
AF245479.1 | - | 13 | 2029 | |||
AK143800.1 | - | 12 | 2208 | |||
JQ437838.1 | - | 13 | 1897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
