siRNA Details
Gene Symbol: Rnase4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021472.4 | NP_067447.2 | 2 | 493 | ||
NM_201239.3 | NP_957691.1 | 2 | 790 | |||
GenBank mRNA | AB041044.1 | - | 2 | 399 | ||
AK050451.1 | - | 2 | 390 | |||
AY762362.1 | - | 2 | 392 | |||
BC005569.1 | - | 2 | 367 | 3967265 | ||
BC061105.1 | - | 2 | 374 | 30296907 | ||
BC066843.1 | - | 2 | 454 | 30059308 |
