siRNA Details
Gene Symbol: Cysltr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281859.1 | NP_001268788.1 | 4 | 942 | ||
NM_001281862.1 | NP_001268791.1 | 2 | 784 | |||
NM_021476.5 | NP_067451.2 | 4 | 961 | |||
GenBank mRNA | AB044087.1 | - | 4 | 872 | ||
AF205830.1 | - | 4 | 750 | |||
AF263370.1 | - | 4 | 862 | |||
AK033476.1 | - | 4 | 745 | |||
BC027102.1 | - | 4 | 765 | 5012266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
