siRNA Details
Gene Symbol: Treh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277847.1 | NP_001264776.1 | 7 | 737 | ||
NM_001277851.1 | NP_001264780.1 | 7 | 741 | |||
NM_021481.3 | NP_067456.1 | 8 | 842 | |||
GenBank mRNA | AF136944.1 | - | 8 | 837 | ||
AK002566.1 | - | 7 | 740 | |||
AK008912.1 | - | 8 | 827 | |||
BC019214.1 | - | 7 | 732 | 5064708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
