siRNA Details
Gene Symbol: Arhgap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021493.2 | NP_067468.2 | 8 | 2188 | ||
GenBank mRNA | BM943900.1 | - | 2 | 354 | ||
CX243267.1 | - | 2 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
