siRNA Details
Gene Symbol: Pvrl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021495.4 | NP_067470.1 | 3 | 974 | ||
NM_021496.3 | NP_067471.1 | 3 | 974 | |||
NM_021497.2 | NP_067472.1 | 3 | 974 | |||
GenBank mRNA | AF195833.1 | - | 3 | 971 | ||
AF195834.1 | - | 3 | 781 | |||
AF195835.1 | - | 3 | 781 | |||
AK011949.1 | - | 3 | 974 | |||
AK013863.1 | - | 60 | ||||
BC125588.1 | - | 3 | 900 | 40130103 | ||
BC132187.1 | - | 3 | 900 | 40130464 | ||
BU701648.1 | - | 2 | 340 |
