siRNA Details
Gene Symbol: Pole3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021498.2 | NP_067473.2 | 3 | 254 | ||
GenBank mRNA | AF230806.1 | - | 3 | 254 | ||
AK007693.1 | - | 449 | ||||
BC024996.1 | - | 3 | 254 | 5253060 | ||
BC034004.1 | - | 262 |
