siRNA Details
Gene Symbol: Trappc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021502.2 | NP_067477.1 | 3 | 255 | ||
GenBank mRNA | AB030200.1 | - | 3 | 248 | ||
AK007526.1 | - | 3 | 225 | |||
BC028502.1 | - | 3 | 235 | 1366538 | ||
BC114968.1 | - | 223 | ||||
BY091059.1 | - | 3 | 256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
