siRNA Details
Gene Symbol: Thap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021513.2 | NP_067488.1 | 1 | 1581 | ||
GenBank mRNA | AB041579.1 | - | 1 | 1542 | ||
AK165981.1 | - | 1 | 1542 | |||
AK166109.1 | - | 1 | 1578 | |||
AK170457.1 | - | 1 | 1553 | |||
BC003949.1 | - | 1 | 1134 | |||
BC138966.1 | - | 1 | 1493 | 8861988 | ||
BC138987.1 | - | 1 | 1493 | 8862009 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
