siRNA Details
Gene Symbol: Huwe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021523.4 | NP_067498.4 | 15, 16 | 1832 | ||
GenBank mRNA | AK031374.1 | - | 16, 17 | 1859 | ||
AK049460.1 | - | 15, 16 | 1788 | |||
AK142956.1 | - | 16, 17 | 1819 | |||
AK163416.1 | - | 16, 17 | 1982 | |||
AK165140.1 | - | 15, 16 | 1768 | |||
AK165648.1 | - | 16, 17 | 1905 | |||
AY772010.3 | - | 15, 16 | 1769 | |||
AY929611.1 | - | 13, 14 | 1379 | |||
DQ097265.1 | - | 15, 16 | 1788 |

More Information
Other Species Targeted

Gene Ontology Categories:
