siRNA Details
Gene Symbol: Huwe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021523.4 | NP_067498.4 | 14, 15 | 1692 | ||
GenBank mRNA | AK031374.1 | - | 15, 16 | 1719 | ||
AK049460.1 | - | 14, 15 | 1648 | |||
AK142956.1 | - | 15, 16 | 1679 | |||
AK163416.1 | - | 15, 16 | 1842 | |||
AK165140.1 | - | 14, 15 | 1628 | |||
AK165648.1 | - | 15, 16 | 1765 | |||
AY772010.3 | - | 14, 15 | 1629 | |||
AY929611.1 | - | 12, 13 | 1239 | |||
DQ097265.1 | - | 14, 15 | 1648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
