siRNA Details
Gene Symbol: Huwe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021523.4 | NP_067498.4 | 23 | 2734 | ||
GenBank mRNA | AK031374.1 | - | 24 | 2761 | ||
AK049460.1 | - | 23 | 2690 | |||
AK142956.1 | - | 25 | 2958 | |||
AK163416.1 | - | 24 | 2884 | |||
AK165140.1 | - | 24 | 2904 | |||
AY772010.3 | - | 23 | 2671 | |||
AY929611.1 | - | 21 | 2281 | |||
DQ097265.1 | - | 23 | 2690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
