siRNA Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021525.2 | NP_067500.1 | 4 | 577 | ||
GenBank mRNA | AJ276895.1 | - | 4 | 447 | ||
AK009709.1 | - | 4 | 577 | |||
AK012585.1 | - | 4 | 534 | |||
AK077751.1 | - | 4 | 530 | |||
AK153377.1 | - | 4 | 507 | |||
AK168153.1 | - | 4 | 559 | |||
BC004574.1 | - | 4 | 501 | 3586632 | ||
BC053404.1 | - | 4 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
