siRNA Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021525.2 | NP_067500.1 | 3 | 425 | ||
GenBank mRNA | AJ276895.1 | - | 3 | 295 | ||
AK009709.1 | - | 3 | 425 | |||
AK012585.1 | - | 3 | 383 | |||
AK077751.1 | - | 3 | 378 | |||
AK153377.1 | - | 3 | 356 | |||
AK168153.1 | - | 3 | 407 | |||
BC004574.1 | - | 3 | 349 | 3586632 | ||
BC053404.1 | - | 3 | 380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
