siRNA Details
Gene Symbol: Ppp2r3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021529.3 | NP_067504.2 | 10, 11 | 1066 | ||
GenBank mRNA | AB041577.1 | - | 10, 11 | 1055 | ||
AJ238247.2 | - | 9, 10 | 893 | |||
AK013403.1 | - | 9, 10 | 862 | |||
AK015746.1 | - | 9, 10 | 895 | |||
AK028063.1 | - | 10, 11 | 1031 | |||
AK028254.1 | - | 9, 10 | 985 | |||
AK033405.1 | - | 10, 11 | 1065 | |||
AK040589.1 | - | 9, 10 | 894 | |||
AK087572.1 | - | 9, 10 | 889 | |||
AK088240.1 | - | 9, 10 | 975 | |||
AK163192.1 | - | 1065 | ||||
AK168298.1 | - | 10, 11 | 1068 | |||
BC024754.1 | - | 10, 11 | 1029 | 5375405 | ||
BC138295.1 | - | 9, 10 | 862 | 8861314 | ||
BC138296.1 | - | 9, 10 | 862 | 8861315 |

More Information
Other Species Targeted

Gene Ontology Categories:
