siRNA Details
Gene Symbol: Ppp2r3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021529.3 | NP_067504.2 | 4, 5 | 518 | ||
GenBank mRNA | AB041577.1 | - | 4, 5 | 507 | ||
AJ238247.2 | - | 3, 4 | 345 | |||
AK013403.1 | - | 3, 4 | 314 | |||
AK015746.1 | - | 3, 4 | 347 | |||
AK028063.1 | - | 4, 5 | 483 | |||
AK028254.1 | - | 3, 4 | 437 | |||
AK033405.1 | - | 4, 5 | 517 | |||
AK040589.1 | - | 3, 4 | 346 | |||
AK087572.1 | - | 3, 4 | 341 | |||
AK088240.1 | - | 3, 4 | 427 | |||
AK163192.1 | - | 517 | ||||
AK168298.1 | - | 4, 5 | 520 | |||
BC024754.1 | - | 4, 5 | 481 | 5375405 | ||
BC138295.1 | - | 3, 4 | 314 | 8861314 | ||
BC138296.1 | - | 3, 4 | 314 | 8861315 |
