siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 7, 8 | 1043 | ||
GenBank mRNA | AF224508.1 | - | 7, 8 | 989 | ||
AK034237.1 | - | 7, 8 | 1045 | |||
AK039404.1 | - | 7, 8 | 1008 | |||
AK171610.1 | - | 6, 7 | 819 | |||
AK173014.1 | - | 7, 8 | 877 | |||
AK209468.1 | - | 2, 3 | 118 | |||
BC030388.1 | - | 7, 8 | 1005 | 5401143 |
