siRNA Details
Gene Symbol: Zfp191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021559.2 | NP_067534.2 | 4 | 856 | ||
GenBank mRNA | AB221566.1 | - | 3 | 715 | ||
AF149093.1 | - | 3 | 715 | |||
AK146726.1 | - | 4 | 854 | |||
AY832930.1 | - | 4 | 895 | |||
BC011345.1 | - | 4 | 899 | 3992743 | ||
BC054832.1 | - | 4 | 856 | 6467818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
