siRNA Details
Gene Symbol: Fxyd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022004.6 | NP_071287.1 | 3 | 391 | ||
GenBank mRNA | AA521976.1 | - | 3 | 130 | ||
AB032010.2 | - | 3 | 104 | |||
AK003888.1 | - | 3 | 154 | |||
AK018354.1 | - | 3 | 124 | |||
BC042579.1 | - | 3 | 137 | 3709336 | ||
BC051127.1 | - | 3 | 162 | 6531451 | ||
BQ891313.1 | - | 3 | 105 | |||
BY704145.1 | - | 3 | 148 | |||
W34202.1 | - | 3 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
