siRNA Details
Gene Symbol: Myg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021713.2 | NP_068359.1 | 5 | 696 | ||
GenBank mRNA | AF289484.1 | - | 5 | 688 | ||
AK002656.1 | - | 5 | 671 | |||
AK004104.1 | - | 5 | 671 | |||
AK013232.1 | - | 5 | 642 | |||
BC023124.1 | - | 4 | 823 | |||
BC028501.1 | - | 5 | 681 | 5256915 |
