siRNA Details
Gene Symbol: Sap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021788.2 | NP_068560.1 | 2 | 638 | ||
GenBank mRNA | AF075136.1 | - | 2 | 649 | ||
AK010928.1 | - | 2 | 639 | |||
AK088745.1 | - | 2 | 643 | |||
BC132081.1 | - | 2 | 621 | 40130358 | ||
BC132087.1 | - | 2 | 621 | 40130364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
