siRNA Details
Gene Symbol: Tmem8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021793.2 | NP_068565.1 | 11 | 1922 | ||
GenBank mRNA | AB045293.1 | - | 11 | 1921 | ||
AK044384.1 | - | 11 | 1923 | |||
AK079893.1 | - | 4 | 1820 | |||
BC005722.1 | - | 10 | 1798 | 3595205 |
