siRNA Details
Gene Symbol: Il21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291041.1 | NP_001277970.1 | 4 | 424 | ||
NM_021782.3 | NP_068554.1 | 4 | 424 | |||
GenBank mRNA | AF254070.1 | - | 4 | 406 | ||
AY428162.1 | - | 4 | 353 | |||
BC125414.1 | - | 4 | 394 | 40129929 | ||
BC125416.1 | - | 4 | 394 | 40129931 | ||
DQ645418.1 | - | 4 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
