siRNA Details
Gene Symbol: Mapk8ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021921.3 | NP_068740.3 | 3 | 749 | ||
GenBank mRNA | AF220195.1 | - | 3 | 468 | ||
AF310135.1 | - | 3 | 530 | |||
AK014339.1 | - | 3 | 453 | |||
AK158968.1 | - | 3 | 603 | |||
BC029704.1 | - | 3 | 528 | 5359447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
