siRNA Details
Gene Symbol: Gmfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022023.2 | NP_071306.2 | 2 | 228 | ||
GenBank mRNA | AB001732.1 | - | 2 | 160 | ||
AB050013.1 | - | 2 | 170 | |||
AF297220.1 | - | 2 | 206 | |||
AK013947.1 | - | 2 | 185 | |||
AK013970.1 | - | 2 | 181 | |||
AK142014.1 | - | 2 | 200 | |||
BC040233.1 | - | 2 | 162 | 5364203 | ||
BC049134.1 | - | 2 | 166 | 6831501 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK034816.1 | - | 397 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
