siRNA Details
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271843.1 | NP_001258772.1 | 2 | 388 | ||
NM_022026.3 | NP_071309.1 | 2 | 388 | |||
GenBank mRNA | AB037180.1 | - | 2 | 185 | ||
AK050042.1 | - | 2 | 388 | |||
AK154151.1 | - | 2 | 392 | |||
BC024105.1 | - | 2 | 359 | 5096952 | ||
CT010174.1 | - | 2 | 182 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK050325.1 | - | 1444 | Chr. - - [-] on Build |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
