siRNA Details
Gene Symbol: Sav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022028.2 | NP_071311.1 | 3 | 806 | ||
GenBank mRNA | AJ292968.1 | - | 3 | 806 | ||
AK006655.1 | - | 2 | 243 | |||
AK034983.1 | - | 1 | 1679 | |||
BC019377.1 | - | 3 | 739 | 4021056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
