siRNA Details
Gene Symbol: Sv2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022030.3 | NP_071313.1 | 11 | 2153 | ||
GenBank mRNA | AK028318.1 | - | 11 | 2098 | ||
AK080502.1 | - | 11 | 2107 | |||
AK080525.1 | - | 11 | 2196 | |||
AK122360.1 | - | 11 | 2189 | |||
AK144166.1 | - | 11 | 2176 | |||
BC026494.1 | - | 5 | 500 | |||
BC046587.1 | - | 11 | 2153 | 6493509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
