siRNA Details
Gene Symbol: Clstn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022319.2 | NP_071714.2 | 16 | 2857 | ||
GenBank mRNA | AJ278069.1 | - | 16 | 2805 | ||
AK048369.1 | - | 9 | 1391 | |||
AK220500.1 | - | 16 | 2871 | |||
BC063058.1 | - | 16 | 2857 | 5705591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
