siRNA Details
Gene Symbol: Clstn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022319.2 | NP_071714.2 | 9 | 1568 | ||
GenBank mRNA | AJ278069.1 | - | 9 | 1516 | ||
AK048369.1 | - | 2 | 102 | |||
AK220500.1 | - | 9 | 1582 | |||
BC063058.1 | - | 9 | 1568 | 5705591 |

More Information
Other Species Targeted

Gene Ontology Categories:
