siRNA Details
Gene Symbol: Ctsm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022326.3 | NP_071721.2 | 5 | 659 | ||
GenBank mRNA | AF202528.1 | - | 5 | 702 | ||
AF456463.1 | - | 4 | 568 | |||
AK005428.1 | - | 5 | 659 | |||
AK005550.1 | - | 5 | 654 | |||
AY014777.1 | - | 5 | 624 |
