siRNA Details
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022327.5 | NP_071722.1 | 2, 3 | 362 | ||
GenBank mRNA | AF174296.1 | - | 330 | |||
AK017698.1 | - | 2, 3 | 208 | |||
AK033193.1 | - | 2, 3 | 217 | |||
AK159993.1 | - | 2, 3 | 307 | |||
AK205791.1 | - | 2, 3 | 176 | |||
BC006907.1 | - | 2, 3 | 206 | 3599947 | ||
BY237628.1 | - | 2, 3 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
