siRNA Details
Gene Symbol: St7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083315.2 | NP_001076784.1 | 9 | 1215 | ||
NM_001289624.1 | NP_001276553.1 | 10 | 1208 | |||
NM_001289625.1 | NP_001276554.1 | 10 | 1208 | |||
NM_001289626.1 | NP_001276555.1 | 9 | 1139 | |||
NM_001289627.1 | NP_001276556.1 | 10 | 942 | |||
NM_001289629.1 | NP_001276558.1 | 9 | 873 | |||
NM_022332.3 | NP_071727.2 | 10 | 1284 | |||
GenBank mRNA | AF234885.1 | - | 9 | 1166 | ||
AK079107.1 | - | 10 | 1285 | |||
AK132103.1 | - | 9 | 1169 | |||
AK151550.1 | - | 9 | 1170 | |||
AY007792.1 | - | 10 | 1170 | |||
AY007793.1 | - | 9 | 1101 | |||
AY007794.1 | - | 11 | 1199 | |||
AY007795.1 | - | 10 | 1130 | |||
AY007796.1 | - | 11 | 1009 | |||
AY007797.1 | - | 10 | 940 | |||
AY007798.1 | - | 10 | 942 | |||
AY007799.1 | - | 9 | 873 | |||
AY551821.1 | - | 10 | 935 | |||
AY551822.1 | - | 9 | 866 | |||
BC012719.1 | - | 4 | 355 | |||
BC024652.1 | - | 10 | 1208 | 3660309 | ||
BC051429.1 | - | 9 | 2894 | |||
BC060630.1 | - | 9 | 2957 | |||
BC108418.1 | - | 10 | 1228 | 30603484 | ||
CT010245.1 | - | 9 | 792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
