siRNA Details
Gene Symbol: Ngb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294308.1 | NP_001281237.1 | 2, 3 | 484 | ||
NM_022414.2 | NP_071859.1 | 2, 3 | 484 | |||
GenBank mRNA | AJ245945.1 | - | 2, 3 | 281 | ||
AK140173.1 | - | 2, 3 | 468 | |||
AK165775.1 | - | 2, 3 | 290 | |||
BC024263.1 | - | 2, 3 | 484 | 5133442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
