siRNA Details
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022417.3 | NP_071862.2 | 3 | 524 | ||
GenBank mRNA | AB030199.1 | - | 3 | 467 | ||
AF272044.1 | - | 3 | 431 | |||
AF282981.1 | - | 3 | 452 | |||
AK036377.1 | - | 3 | 470 | |||
AK156930.1 | - | 3 | 427 | |||
BC012952.1 | - | 3 | 402 | 2648812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
