siRNA Details
Gene Symbol: Svs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021363.2 | NP_067338.2 | 3 | 1121 | ||
GenBank mRNA | AF242220.1 | - | 3 | 1106 | ||
AK020575.1 | - | 3 | 1118 | |||
DV048837.1 | - | 2 | 689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
