siRNA Details
Gene Symbol: Fcamr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170632.1 | NP_001164103.1 | 7 | 1755 | ||
NM_144960.2 | NP_659209.2 | 7 | 1587 | |||
GenBank mRNA | AB048834.1 | - | 7 | 1599 | ||
BC110549.1 | - | 7 | 1758 | 40047147 | ||
BC110550.1 | - | 7 | 1590 | 40047153 | ||
FJ011703.1 | - | 6 | 1668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
