siRNA Details
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022565.2 | NP_072087.2 | 6 | 2136 | ||
GenBank mRNA | AB036838.1 | - | 5 | 1586 | ||
AK140992.1 | - | 6 | 2137 |

More Information
Other Species Targeted

Gene Ontology Categories:
