siRNA Details
Gene Symbol: Mrps22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025485.3 | NP_079761.1 | 5, 6 | 772 | ||
GenBank mRNA | AK003836.1 | - | 4, 5 | 540 | ||
AK013925.1 | - | 5, 6 | 753 | |||
AK154600.1 | - | 5, 6 | 753 | |||
BC051198.1 | - | 5, 6 | 752 | 1396237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
