siRNA Details
Gene Symbol: Mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026080.2 | NP_080356.1 | 4 | 313 | ||
GenBank mRNA | AK003117.1 | - | 4 | 297 | ||
AK007338.1 | - | 4 | 297 | |||
AK010590.1 | - | 4 | 294 | |||
AK012461.1 | - | 4 | 302 | |||
AK014099.1 | - | 4 | 300 | |||
BC029262.1 | - | 4 | 287 | 4911699 | ||
BY100364.1 | - | 4 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
