siRNA Details
Gene Symbol: Htra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019752.3 | NP_062726.3 | 3 | 1040 | ||
GenBank mRNA | AF164513.1 | - | 3 | 919 | ||
AF175324.1 | - | 3 | 936 | |||
AK011536.1 | - | 1846 | ||||
AK146338.1 | - | 3 | 931 | |||
AK159188.1 | - | 3 | 860 | |||
AK176936.1 | - | 1 | 23 | |||
AK178298.1 | - | 1 | 23 | |||
AK182973.1 | - | 1 | 23 | |||
AK186135.1 | - | 1 | 23 | |||
AK188221.1 | - | 1 | 23 | |||
AK215600.1 | - | 1 | 23 | |||
BC033276.1 | - | 3 | 900 | |||
BC049880.1 | - | 3 | 1040 | 5321861 |

More Information
Other Species Targeted

Gene Ontology Categories:
