siRNA Details
Gene Symbol: Rpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022891.3 | NP_075029.1 | 4 | 324 | ||
GenBank mRNA | AF287271.1 | - | 4 | 258 | ||
AK002579.1 | - | 4 | 270 | |||
AK010680.1 | - | 4 | 273 | |||
AK012694.1 | - | 4 | 269 | |||
AK018730.1 | - | 4 | 269 | |||
AK150671.1 | - | 4 | 269 | |||
AK168133.1 | - | 4 | 255 | |||
BC025918.1 | - | 4 | 258 | 5252035 | ||
BC081448.1 | - | 4 | 258 | 5714104 | ||
CN840510.1 | - | 4 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
