siRNA Details
Gene Symbol: Zfp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022981.4 | NP_075357.4 | 8 | 2114 | ||
GenBank mRNA | AJ242914.1 | - | 8 | 1869 | ||
AK013583.1 | - | 8 | 1920 | |||
AK040989.1 | - | 8 | 1848 | |||
BC006657.1 | - | 8 | 1863 | 3500984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
